Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007378.1 | NP_031404.1 | 26 - 27 | 3949 | 59 | |
GenBank mRNA | AF000149.1 | - | 26 - 27 | 3949 | 59 | |
AK052916.1 | - | 3 - 4 | 307 | 59 | ||
BC057853.1 | - | 26 - 27 | 3949 | 6492139 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |