Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291809.1 | NP_001278738.1 | 8 - 9 | 1250 | 91 | |
NM_007882.3 | NP_031908.3 | 8 - 9 | 1250 | 91 | ||
GenBank mRNA | BC144845.1 | - | 8 - 9 | 1235 | 91 | |
Y11169.1 | - | 8 - 9 | 1250 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |