Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042623.2 | NP_001036088.1 | 9 - 10 | 2217 | 125 | |
NM_001271579.1 | NP_001258508.1 | 10 - 11 | 2344 | 125 | ||
NM_007905.3 | NP_031931.2 | 10 - 11 | 2373 | 125 | ||
NR_073364.1 | 10 - 11 | 2380 | 125 | |||
GenBank mRNA | AK036370.1 | - | 10 - 11 | 2333 | 125 | |
AK132756.1 | - | 9 - 10 | 2175 | 125 | ||
AK167590.1 | - | 9 - 10 | 2206 | 125 | ||
BC046535.1 | - | 10 - 11 | 2360 | 4935713 | 125 | |
BC052394.1 | - | 10 - 11 | 2706 | 5719837 | 125 | |
S73882.1 | - | 10 - 11 | 2335 | 125 | ||
S73883.1 | - | 10 - 11 | 2342 | 125 | ||
S73884.1 | - | 9 - 10 | 2583 | 125 | ||
U63386.1 | - | 9 - 10 | 2301 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |