Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008130.2 | NP_032156.2 | 14 - 15 | 2867 | 62 | |
GenBank mRNA | BC141135.1 | - | 14 - 15 | 2568 | 9055747 | 62 |
BC145445.1 | - | 14 - 15 | 2568 | 9054003 | 62 | |
CA752350.1 | - | 3 - 4 | 725 | 62 | ||
X95255.1 | - | 14 - 15 | 2607 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |