Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008136.2 | NP_032162.2 | 3 - 4 | 592 | 92 | |
GenBank mRNA | BC010298.1 | - | 3 - 4 | 547 | 3155411 | 92 |
BC013243.1 | - | 3 - 4 | 616 | 92 | ||
BC129969.1 | - | 1 - 2 | 124 | 92 | ||
BC129970.1 | - | 1 - 2 | 124 | 92 | ||
BM949933.1 | - | 3 - 4 | 338 | 92 | ||
X65026.1 | - | 2 - 3 | 235 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |