Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008326.1 | NP_032352.1 | 1 - 2 | 250 | 84 | |
GenBank mRNA | AK002545.1 | - | 1 - 2 | 272 | 84 | |
AK171743.1 | - | 1 - 2 | 231 | 84 | ||
BC145957.1 | - | 1 - 2 | 62 | 40131260 | 84 | |
U19119.1 | - | 1 - 2 | 250 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |