Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008446.2 | NP_032472.2 | 1 - 2 | 522 | 142 | |
GenBank mRNA | AK084837.1 | - | 1 - 2 | 499 | 142 | |
AK135179.1 | - | 1 - 2 | 470 | 142 | ||
AK171938.1 | - | 1 - 2 | 436 | 142 | ||
BC050946.1 | - | 1 - 2 | 460 | 6822283 | 142 | |
BC054537.1 | - | 1 - 2 | 444 | 142 | ||
CF742002.1 | - | 1 - 2 | 522 | 142 | ||
D12646.1 | - | 1 - 2 | 849 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |