Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277928.1 | NP_001264857.1 | 19 - 20 | 3041 | 105 | |
NM_008484.2 | NP_032510.2 | 19 - 20 | 3080 | 105 | ||
GenBank mRNA | AK078970.1 | - | 4 - 5 | 596 | 105 | |
AK155769.1 | - | 3893 | 105 | |||
BC008516.1 | - | 19 - 20 | 3041 | 3592888 | 105 | |
U43298.1 | - | 19 - 20 | 3056 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |