Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK028796.1 | - | 2 - 3 | 397 | 94 | |
AK171128.1 | - | 2 - 3 | 341 | 94 | ||
U89992.2 | - | 162 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus 1700 Inventoried Probe spans exons |