Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281980.1 | NP_001268909.1 | 17 - 18 | 1438 | 91 | |
NM_001281981.1 | NP_001268910.1 | 18 - 19 | 1479 | 91 | ||
NM_008511.3 | NP_032537.3 | 17 - 18 | 1710 | 91 | ||
GenBank mRNA | AK153865.1 | - | 17 - 18 | 1440 | 91 | |
AK160009.1 | - | 17 - 18 | 1670 | 91 | ||
AK166635.1 | - | 15 - 16 | 1159 | 91 | ||
AK168775.1 | - | 15 - 16 | 1136 | 91 | ||
AK169578.1 | - | 18 - 19 | 1463 | 91 | ||
AY753324.1 | - | 16 - 17 | 1273 | 91 | ||
BC052909.1 | - | 17 - 18 | 1646 | 30076553 | 91 | |
U10484.1 | - | 17 - 18 | 1416 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |