Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tulp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF105712.1 | - | 10 - 11 | 1289 | 113 | |
AK054304.1 | - | 7 - 8 | 1267 | 113 | ||
AK077200.1 | - | 10 - 11 | 1298 | 113 | ||
AK077211.1 | - | 7 - 8 | 1095 | 113 | ||
AK161165.1 | - | 7 - 8 | 1246 | 113 | ||
BC048740.1 | - | 3 - 4 | 428 | 113 | ||
BC089545.1 | - | 7 - 8 | 1094 | 6770730 | 113 | |
X69827.1 | - | 4 - 5 | 551 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |