Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161774.2 | NP_001155246.1 | 3 - 4 | 407 | 56 | |
NM_001285520.1 | NP_001272449.1 | 3 - 4 | 737 | 56 | ||
NM_009176.5 | NP_033202.3 | 3 - 4 | 751 | 56 | ||
GenBank mRNA | AK005053.1 | - | 3 - 4 | 696 | 56 | |
AK136442.1 | - | 3 - 4 | 688 | 56 | ||
BC006710.1 | - | 3 - 4 | 375 | 3584323 | 56 | |
CX240214.1 | - | 3 - 4 | 367 | 56 | ||
X84234.1 | - | 3 - 4 | 230 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |