Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161774.2 | NP_001155246.1 | 8 - 9 | 791 | 80 | |
NM_001285520.1 | NP_001272449.1 | 8 - 9 | 1121 | 80 | ||
NM_001285521.1 | NP_001272450.1 | 7 - 8 | 743 | 80 | ||
NM_009176.5 | NP_033202.3 | 8 - 9 | 1135 | 80 | ||
GenBank mRNA | AK005053.1 | - | 8 - 9 | 1080 | 80 | |
AK013016.1 | - | 7 - 8 | 682 | 80 | ||
AK136442.1 | - | 8 - 9 | 1072 | 80 | ||
BC006710.1 | - | 8 - 9 | 759 | 3584323 | 80 | |
X84234.1 | - | 8 - 9 | 614 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |