Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tapbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025313.1 | NP_001020484.1 | 5 - 6 | 1430 | 87 | |
NM_009318.2 | NP_033344.1 | 5 - 6 | 1430 | 87 | ||
GenBank mRNA | AF043943.1 | - | 5 - 6 | 1288 | 87 | |
AF106278.1 | - | 5 - 6 | 1220 | 87 | ||
AJ316613.2 | - | 5 - 6 | 1220 | 87 | ||
AK014566.1 | - | 5 - 6 | 1285 | 87 | ||
AK054174.1 | - | 2 - 3 | 605 | 87 | ||
AK077299.1 | - | 5 - 6 | 1254 | 87 | ||
AK151134.1 | - | 5 - 6 | 1251 | 87 | ||
AK151807.1 | - | 5 - 6 | 1233 | 87 | ||
AK151878.1 | - | 5 - 6 | 1247 | 87 | ||
AK152019.1 | - | 5 - 6 | 1246 | 87 | ||
AK152726.1 | - | 2 - 3 | 699 | 87 | ||
AK169697.1 | - | 5 - 6 | 1251 | 87 | ||
AK170528.1 | - | 5 - 6 | 1241 | 87 | ||
AY734468.1 | - | 5 - 6 | 1204 | 87 | ||
AY734469.1 | - | 5 - 6 | 1203 | 87 | ||
AY734471.1 | - | 5 - 6 | 1218 | 87 | ||
AY734475.1 | - | 4 - 5 | 1186 | 87 | ||
AY734476.1 | - | 1 - 2 | 171 | 87 | ||
BC013236.1 | - | 4 - 5 | 1028 | 87 | ||
BC015074.1 | - | 5 - 6 | 1243 | 4223922 | 87 | |
KJ650208.1 | - | 5 - 6 | 1220 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |