Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31113241 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31113236 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31113240 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31113238 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31113239 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31113237 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1954998 |
BC145595.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1954998 |
BC145596.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1954998 |
U81490.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1954998 |
AK155268.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1954998 |
BC015077.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P220260 |
U81490.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P220260 |
AK155268.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P220260 |
AK204879.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P220260 |
AK205199.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P220260 |
AK139088.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P220260 |
U81490.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P220260 |
AK155268.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P220260 |
AK204879.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P220260 |
AK205199.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P220260 |
AK139088.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
260908 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
806529 |
XM_001479595.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
806529 |
XM_001479611.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
806529 |
XM_001480937.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
806529 |
XM_896600.2 |
refseq_mrna_mouse_XM |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P174142 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P220260 |
U81490.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P220260 |
AK155268.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P220260 |
AK204879.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P220260 |
AK205199.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P220260 |
AK139088.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P220260 |
U81490.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P220260 |
AK155268.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P220260 |
AK204879.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P220260 |
AK205199.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P220260 |
AK139088.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1954998 |
BC145595.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1954998 |
BC145596.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1954998 |
U81490.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1954998 |
AK155268.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1954998 |
BC015077.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P220260 |
U81490.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P220260 |
AK155268.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P220260 |
AK204879.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P220260 |
AK205199.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P220260 |
AK139088.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P220260 |
U81490.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P220260 |
AK155268.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P220260 |
AK204879.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P220260 |
AK205199.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P220260 |
AK139088.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113281 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113275 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113234 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113236 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113247 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113249 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113262 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113261 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113241 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113239 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113244 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113245 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113274 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113235 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113254 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113246 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113276 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113260 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113278 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113259 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113242 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113232 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113238 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113252 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113237 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31113257 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P174142 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P220260 |
U81490.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P220260 |
AK155268.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P220260 |
AK204879.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P220260 |
AK205199.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P220260 |
AK139088.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P220260 |
U81490.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P220260 |
AK155268.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P220260 |
AK204879.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P220260 |
AK205199.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P220260 |
AK139088.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P104096 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P122538 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P129120 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P137696 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P139004 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P174142 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13468810 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13468806 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13468795 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13468825 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13468776 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13468790 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13468820 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31113238 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31113240 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31113237 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31113241 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31113236 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31113239 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113185 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113275 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113333 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113303 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113315 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113347 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113297 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113332 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113314 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113292 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113300 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113308 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113254 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113247 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113242 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113322 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113344 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113296 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113318 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113338 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113323 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113252 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113266 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113269 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113260 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113356 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113295 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113330 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113320 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113361 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113301 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113352 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113359 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113263 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113283 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113286 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113349 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113348 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113241 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113327 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113282 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113343 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113285 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113183 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113272 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113325 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113354 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113345 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113238 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113312 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113307 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113289 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113244 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113281 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113257 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113236 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113249 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113305 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113341 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113278 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113357 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113353 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113276 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113351 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113335 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113264 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31113336 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113260 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113254 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113241 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113259 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113274 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113262 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13468806 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113278 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113261 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113236 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113272 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113276 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13468790 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113263 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13468804 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113240 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113275 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113264 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113249 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13468788 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113266 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113244 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13468821 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113235 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113269 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113281 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113234 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13468807 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113267 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31113242 |
- |
- |