Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009462.1 | NP_033488.1 | 4 - 5 | 1243 | 81 | |
GenBank mRNA | AK077453.1 | - | 4 - 5 | 1265 | 81 | |
AK129083.1 | - | 4 - 5 | 1251 | 81 | ||
AK137015.1 | - | 4 - 5 | 1265 | 81 | ||
AK153675.1 | - | 4 - 5 | 1735 | 81 | ||
AK157970.1 | - | 4 - 5 | 1267 | 81 | ||
AK165156.1 | - | 4 - 5 | 1263 | 81 | ||
AK166243.1 | - | 4 - 5 | 1260 | 81 | ||
AK172443.1 | - | 4 - 5 | 1265 | 81 | ||
BC007134.1 | - | 4 - 5 | 1245 | 3491297 | 81 | |
D84096.1 | - | 4 - 5 | 1243 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |