Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009577.3 | NP_033603.2 | 1 - 2 | 238 | 63 | |
GenBank mRNA | AK146692.1 | - | 1 - 2 | 232 | 63 | |
AK159077.1 | - | 1 - 2 | 224 | 63 | ||
BC050942.1 | - | 1 - 2 | 196 | 6821378 | 63 | |
BC067196.1 | - | 1 - 2 | 196 | 6821378 | 63 | |
BY026558.1 | - | 1 - 2 | 252 | 63 | ||
U69133.1 | - | 1 - 2 | 236 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |