Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278226.1 | NP_001265155.1 | 5 - 6 | 501 | 90 | |
NM_010059.3 | NP_034189.1 | 5 - 6 | 501 | 90 | ||
GenBank mRNA | AK076777.1 | - | 5 - 6 | 492 | 90 | |
BC116767.1 | - | 5 - 6 | 386 | 40126086 | 90 | |
BC119081.1 | - | 5 - 6 | 386 | 8733830 | 90 | |
D58419.1 | - | 5 - 6 | 486 | 90 | ||
D64107.1 | - | 5 - 6 | 475 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |