Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191026.1 | NP_001177955.1 | 5 - 6 | 645 | 56 | |
NM_010063.4 | NP_034193.2 | 5 - 6 | 678 | 56 | ||
GenBank mRNA | AF063229.1 | - | 5 - 6 | 512 | 56 | |
AK158589.1 | - | 5 - 6 | 646 | 56 | ||
BC051992.1 | - | 5 - 6 | 406 | 5696629 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |