Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159543.1 | NP_001153015.1 | 6 - 7 | 842 | 105 | |
NM_010074.3 | NP_034204.1 | 6 - 7 | 842 | 105 | ||
GenBank mRNA | AK040409.1 | - | 5 - 6 | 548 | 105 | |
AK085370.1 | - | 6 - 7 | 425 | 105 | ||
AK153939.1 | - | 6 - 7 | 456 | 105 | ||
AK163085.1 | - | 6 - 7 | 847 | 105 | ||
AK164775.1 | - | 6 - 7 | 425 | 105 | ||
BC022183.1 | - | 6 - 7 | 704 | 3982742 | 105 | |
X58384.1 | - | 6 - 7 | 519 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |