Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010267.3 | NP_034397.1 | 2 - 3 | 412 | 123 | |
GenBank mRNA | AK020988.1 | - | 2 - 3 | 396 | 123 | |
AK045460.1 | - | 2 - 3 | 412 | 123 | ||
AK049655.1 | - | 2 - 3 | 382 | 123 | ||
AK083814.1 | - | 2 - 3 | 413 | 123 | ||
AK139057.1 | - | 2 - 3 | 255 | 123 | ||
AK149281.1 | - | 2 - 3 | 423 | 123 | ||
BC048177.1 | - | 2 - 3 | 388 | 6825756 | 123 | |
BC051135.1 | - | 2 - 3 | 377 | 6532839 | 123 | |
BC060212.1 | - | 2 - 3 | 385 | 123 | ||
Y17850.1 | - | 2 - 3 | 400 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |