Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010267.3 | NP_034397.1 | 4 - 5 | 682 | 113 | |
GenBank mRNA | AK020988.1 | - | 4 - 5 | 666 | 113 | |
AK045460.1 | - | 4 - 5 | 682 | 113 | ||
AK083814.1 | - | 4 - 5 | 683 | 113 | ||
AK139057.1 | - | 4 - 5 | 525 | 113 | ||
AK149281.1 | - | 4 - 5 | 693 | 113 | ||
BC048177.1 | - | 4 - 5 | 658 | 6825756 | 113 | |
BC051135.1 | - | 4 - 5 | 647 | 6532839 | 113 | |
BC060212.1 | - | 4 - 5 | 655 | 113 | ||
Y17850.1 | - | 4 - 5 | 670 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |