Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010331.2 | NP_034461.1 | 6 - 7 | 905 | 100 | |
GenBank mRNA | AB002136.1 | - | 6 - 7 | 834 | 100 | |
AB006970.1 | - | 6 - 7 | 841 | 100 | ||
AK077518.1 | - | 6 - 7 | 905 | 100 | ||
AK164219.1 | - | 6 - 7 | 851 | 100 | ||
AK164978.1 | - | 6 - 7 | 905 | 100 | ||
AK168057.1 | - | 6 - 7 | 1108 | 100 | ||
BC006697.1 | - | 6 - 7 | 844 | 3498573 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |