Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010581.3 | NP_034711.1 | 1 - 2 | 176 | 108 | |
GenBank mRNA | AB012693.1 | - | 1 - 2 | 115 | 108 | |
AK151922.1 | - | 1 - 2 | 122 | 108 | ||
BC012667.1 | - | 1 - 2 | 110 | 4187965 | 108 | |
HQ585874.1 | - | 1 - 2 | 43 | 108 | ||
Z25524.1 | - | 1 - 2 | 176 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |