Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010630.2 | NP_034760.2 | 4 - 5 | 495 | 128 | |
GenBank mRNA | AK144304.1 | - | 5 - 6 | 654 | 128 | |
D49545.1 | - | 4 - 5 | 571 | 128 | ||
U92949.1 | - | 4 - 5 | 485 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |