Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Laptm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010686.3 | NP_034816.1 | 4 - 5 | 399 | 73 | |
GenBank mRNA | AF364050.1 | - | 4 - 5 | 417 | 73 | |
AK031802.1 | - | 4 - 5 | 413 | 73 | ||
AK050633.1 | - | 4 - 5 | 414 | 73 | ||
AK089432.1 | - | 4 - 5 | 449 | 73 | ||
AK089508.1 | - | 4 - 5 | 422 | 73 | ||
AK089535.1 | - | 4 - 5 | 414 | 73 | ||
AK150299.1 | - | 4 - 5 | 410 | 73 | ||
AK152918.1 | - | 4 - 5 | 410 | 73 | ||
AK153877.1 | - | 4 - 5 | 413 | 73 | ||
AK155744.1 | - | 4 - 5 | 407 | 73 | ||
AK163146.1 | - | 4 - 5 | 414 | 73 | ||
AK170438.1 | - | 4 - 5 | 401 | 73 | ||
AK170690.1 | - | 4 - 5 | 433 | 73 | ||
AK171053.1 | - | 4 - 5 | 424 | 73 | ||
AK171057.1 | - | 4 - 5 | 449 | 73 | ||
AK171105.1 | - | 4 - 5 | 414 | 73 | ||
AK171116.1 | - | 4 - 5 | 414 | 73 | ||
AK171149.1 | - | 4 - 5 | 422 | 73 | ||
AK171172.1 | - | 414 | 73 | |||
AK171193.1 | - | 4 - 5 | 414 | 73 | ||
AK171213.1 | - | 4 - 5 | 414 | 73 | ||
AK171236.1 | - | 4 - 5 | 414 | 73 | ||
AK171244.1 | - | 4 - 5 | 414 | 73 | ||
AK171250.1 | - | 4 - 5 | 435 | 73 | ||
AK171269.1 | - | 4 - 5 | 451 | 73 | ||
AK171270.1 | - | 4 - 5 | 417 | 73 | ||
AK171274.1 | - | 4 - 5 | 407 | 73 | ||
AK171280.1 | - | 4 - 5 | 447 | 73 | ||
AK171285.1 | - | 4 - 5 | 451 | 73 | ||
AK171481.1 | - | 4 - 5 | 450 | 73 | ||
AK171547.1 | - | 4 - 5 | 414 | 73 | ||
AK171562.1 | - | 4 - 5 | 405 | 73 | ||
AK171571.1 | - | 4 - 5 | 431 | 73 | ||
AK171575.1 | - | 4 - 5 | 449 | 73 | ||
AK172306.1 | - | 4 - 5 | 414 | 73 | ||
AK172486.1 | - | 4 - 5 | 449 | 73 | ||
AK172707.1 | - | 4 - 5 | 414 | 73 | ||
AK172717.1 | - | 1 - 2 | 117 | 73 | ||
AK172885.1 | - | 4 - 5 | 389 | 73 | ||
BC020993.1 | - | 4 - 5 | 399 | 4037686 | 73 | |
BC055057.1 | - | 4 - 5 | 386 | 6529760 | 73 | |
BC114403.1 | - | 4 - 5 | 379 | 73 | ||
BC158032.1 | - | 4 - 5 | 595 | 9007335 | 73 | |
U51239.1 | - | 4 - 5 | 438 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |