Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159577.1 | NP_001153049.1 | 6 - 7 | 1562 | 55 | |
NM_001159578.1 | NP_001153050.1 | 7 - 8 | 1707 | 55 | ||
NM_001159579.1 | NP_001153051.1 | 5 - 6 | 1338 | 55 | ||
NM_001159580.1 | NP_001153052.1 | 5 - 6 | 1308 | 55 | ||
NM_010727.4 | NP_034857.3 | 5 - 6 | 1338 | 55 | ||
GenBank mRNA | AF034745.1 | - | 6 - 7 | 1562 | 55 | |
AF034746.1 | - | 5 - 6 | 1266 | 55 | ||
AK044127.1 | - | 5 - 6 | 1291 | 55 | ||
AK138326.1 | - | 5 - 6 | 1309 | 55 | ||
AK161194.1 | - | 7 - 8 | 1693 | 55 | ||
BC139771.1 | - | 5 - 6 | 1266 | 40111506 | 55 | |
KJ418422.1 | - | 5 - 6 | 1036 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |