Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011019.3 | NP_035149.2 | 3 - 4 | 372 | 118 | |
GenBank mRNA | AB015978.1 | - | 3 - 4 | 1026 | 118 | |
AF058805.1 | - | 3 - 4 | 342 | 118 | ||
AK040020.1 | - | 3 - 4 | 316 | 118 | ||
AK087179.1 | - | 3 - 4 | 328 | 118 | ||
BC137703.1 | - | 2 - 3 | 267 | 8860722 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |