Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011019.3 | NP_035149.2 | 13 - 14 | 1987 | 91 | |
GenBank mRNA | AB015978.1 | - | 13 - 14 | 2638 | 91 | |
AF058805.1 | - | 13 - 14 | 1957 | 91 | ||
AK157508.1 | - | 8 - 9 | 1160 | 91 | ||
BC137703.1 | - | 12 - 13 | 1885 | 8860722 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |