Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011020.3 | NP_035150.3 | 16 - 17 | 2105 | 88 | |
GenBank mRNA | AB001926.1 | - | 16 - 17 | 2071 | 88 | |
AK033950.1 | - | 3 - 4 | 334 | 88 | ||
AK144225.1 | - | 16 - 17 | 2142 | 88 | ||
AK161429.1 | - | 3 - 4 | 251 | 88 | ||
BC012712.1 | - | 16 - 17 | 2174 | 4190161 | 88 | |
BC057002.1 | - | 16 - 17 | 2105 | 5702602 | 88 | |
BC110662.1 | - | 16 - 17 | 2125 | 4511154 | 88 | |
D49482.1 | - | 16 - 17 | 2099 | 88 | ||
U23921.1 | - | 16 - 17 | 2222 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |