Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040435.2 | NP_001035525.1 | 4 - 5 | 948 | 67 | |
GenBank mRNA | AF093542.1 | - | 4 - 5 | 449 | 67 | |
AF156934.1 | - | 4 - 5 | 499 | 67 | ||
AF203445.1 | - | 4 - 5 | 449 | 67 | ||
AF247674.1 | - | 4 - 5 | 449 | 67 | ||
AK076282.1 | - | 971 | 67 | |||
AK145556.1 | - | 4 - 5 | 509 | 67 | ||
AK160291.1 | - | 4 - 5 | 510 | 67 | ||
AK210694.1 | - | 1 - 2 | 75 | 67 | ||
BC003252.1 | - | 4 - 5 | 484 | 3500413 | 67 | |
BC055071.1 | - | 2924 | 67 | |||
BC066184.1 | - | 4 - 5 | 484 | 30457053 | 67 | |
BC068314.1 | - | 4 - 5 | 484 | 30461783 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |