Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040435.2 | NP_001035525.1 | 5 - 6 | 1545 | 85 | |
GenBank mRNA | AF093542.1 | - | 6 - 7 | 758 | 85 | |
AF156934.1 | - | 5 - 6 | 1096 | 85 | ||
AF247674.1 | - | 6 - 7 | 830 | 85 | ||
AK076282.1 | - | 1572 | 85 | |||
AK145556.1 | - | 5 - 6 | 1106 | 85 | ||
AK160291.1 | - | 5 - 6 | 1108 | 85 | ||
BC003252.1 | - | 5 - 6 | 1081 | 3500413 | 85 | |
BC055071.1 | - | 3521 | 85 | |||
BC066184.1 | - | 5 - 6 | 1081 | 30457053 | 85 | |
BC068314.1 | - | 5 - 6 | 1081 | 30461783 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |