Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011543.4 | NP_035673.3 | 1 - 2 | 123 | 63 | |
GenBank mRNA | AF083214.1 | - | 1 - 2 | 47 | 63 | |
AK002983.1 | - | 1 - 2 | 94 | 63 | ||
AK010628.1 | - | 1 - 2 | 109 | 63 | ||
AK012498.1 | - | 1 - 2 | 82 | 63 | ||
AK014245.1 | - | 1 - 2 | 82 | 63 | ||
AK027909.1 | - | 1 - 2 | 81 | 63 | ||
AK078330.1 | - | 1 - 2 | 109 | 63 | ||
AK088339.1 | - | 1 - 2 | 82 | 63 | ||
AK152609.1 | - | 1 - 2 | 81 | 63 | ||
AK160198.1 | - | 1 - 2 | 82 | 63 | ||
AK166673.1 | - | 1 - 2 | 82 | 63 | ||
BC002115.1 | - | 1 - 2 | 66 | 3490159 | 63 | |
BU841654.1 | - | 1 - 2 | 126 | 63 | ||
Z47088.1 | - | 1 - 2 | 72 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |