Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vnn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011704.3 | NP_035834.2 | 1 - 2 | 409 | 72 | |
GenBank mRNA | AJ132098.1 | - | 1 - 2 | 242 | 72 | |
AK145984.1 | - | 1 - 2 | 315 | 72 | ||
AK167427.1 | - | 1 - 2 | 241 | 72 | ||
BB838710.1 | - | 1 - 2 | 411 | 72 | ||
BC019203.1 | - | 1 - 2 | 235 | 5041670 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |