Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zyx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289617.1 | NP_001276546.1 | 6 - 7 | 1426 | 73 | |
NM_001289618.1 | NP_001276547.1 | 5 - 6 | 1333 | 73 | ||
NM_001289619.1 | NP_001276548.1 | 5 - 6 | 1333 | 73 | ||
NM_011777.3 | NP_035907.1 | 6 - 7 | 1426 | 73 | ||
GenBank mRNA | AK031415.1 | - | 6 - 7 | 1224 | 73 | |
AK035743.1 | - | 6 - 7 | 1197 | 73 | ||
AK076491.1 | - | 6 - 7 | 1197 | 73 | ||
AK090110.1 | - | 6 - 7 | 1193 | 73 | ||
AK147812.1 | - | 6 - 7 | 1427 | 73 | ||
AK161709.1 | - | 5 - 6 | 1128 | 73 | ||
AK165290.1 | - | 4 - 5 | 1267 | 73 | ||
AK190613.1 | - | 2 - 3 | 256 | 74 | ||
BC054775.1 | - | 5 - 6 | 1292 | 6808149 | 73 | |
X99063.1 | - | 6 - 7 | 1384 | 73 | ||
Y07711.1 | - | 5 - 6 | 1134 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |