Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Decr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011933.2 | NP_036063.1 | 1 - 2 | 202 | 78 | |
GenBank mRNA | AF155575.1 | - | 1 - 2 | 202 | 78 | |
AK137212.1 | - | 1 - 2 | 133 | 78 | ||
AK149270.1 | - | 1 - 2 | 327 | 78 | ||
AK162961.1 | - | 1 - 2 | 338 | 78 | ||
BC021865.1 | - | 1 - 2 | 174 | 5123899 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |