Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012033.3 | NP_036163.3 | 8 - 9 | 1206 | 152 | |
GenBank mRNA | AF153366.1 | - | 8 - 9 | 1148 | 152 | |
BC010745.1 | - | 8 - 9 | 1151 | 4239192 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |