Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291804.1 | NP_001278733.1 | 8 - 9 | 1284 | 101 | |
NM_013504.5 | NP_038532.3 | 8 - 9 | 1284 | 101 | ||
GenBank mRNA | AK036893.1 | - | 8 - 9 | 1170 | 101 | |
AK079441.1 | - | 8 - 9 | 1286 | 101 | ||
AK081826.1 | - | 8 - 9 | 1192 | 101 | ||
BC109161.1 | - | 8 - 9 | 1121 | 40047883 | 101 | |
BC109162.1 | - | 8 - 9 | 1121 | 101 | ||
X97986.1 | - | 8 - 9 | 1218 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |