Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083319.1 | NP_001076788.1 | 3 - 4 | 681 | 67 | |
NM_013699.2 | NP_038727.1 | 3 - 4 | 681 | 67 | ||
GenBank mRNA | AB221581.1 | - | 2 - 3 | 270 | 67 | |
AK143282.1 | - | 3 - 4 | 682 | 67 | ||
AY182062.1 | - | 2 - 3 | 270 | 67 | ||
BC034749.1 | - | 3 - 4 | 370 | 67 | ||
BC139828.1 | - | 2 - 3 | 298 | 40142981 | 67 | |
BC141082.1 | - | 2 - 3 | 360 | 9055694 | 67 | |
U20086.1 | - | 2 - 3 | 272 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |