Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tjp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282095.1 | NP_001269024.1 | 11 - 12 | 1376 | 66 | |
NM_001282096.1 | NP_001269025.1 | 11 - 12 | 1439 | 66 | ||
NM_013769.3 | NP_038797.2 | 11 - 12 | 1478 | 66 | ||
GenBank mRNA | AF157006.1 | - | 11 - 12 | 1446 | 66 | |
AK029083.1 | - | 11 - 12 | 1425 | 66 | ||
AK134100.1 | - | 11 - 12 | 1479 | 66 | ||
AK167587.1 | - | 11 - 12 | 1377 | 66 | ||
BC012518.1 | - | 11 - 12 | 1443 | 3600848 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |