Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016682.2 | NP_057891.1 | 1 - 2 | 154 | 65 | |
GenBank mRNA | AK146925.1 | - | 1 - 2 | 200 | 65 | |
AK151765.1 | - | 1 - 2 | 159 | 65 | ||
AK152415.1 | - | 1 - 2 | 178 | 65 | ||
AK152977.1 | - | 1 - 2 | 159 | 65 | ||
AK163451.1 | - | 1 - 2 | 150 | 65 | ||
AK164826.1 | - | 1 - 2 | 170 | 65 | ||
AK165321.1 | - | 1 - 2 | 176 | 65 | ||
AK166133.1 | - | 1 - 2 | 186 | 65 | ||
AK168673.1 | - | 1 - 2 | 166 | 65 | ||
AK169168.1 | - | 1 - 2 | 150 | 65 | ||
AK178317.1 | - | 1 - 2 | 62 | 65 | ||
AK184258.1 | - | 1 - 2 | 62 | 65 | ||
AK189465.1 | - | 1 - 2 | 62 | 65 | ||
AK192304.1 | - | 1 - 2 | 62 | 65 | ||
AK194855.1 | - | 1 - 2 | 62 | 65 | ||
AK204958.1 | - | 1 - 2 | 62 | 65 | ||
AK213601.1 | - | 1 - 2 | 62 | 65 | ||
AK219234.1 | - | 1 - 2 | 62 | 65 | ||
AK219985.1 | - | 1 - 2 | 62 | 65 | ||
BC054768.1 | - | 1 - 2 | 180 | 6415071 | 65 | |
U35833.1 | - | 1 - 2 | 155 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |