Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016753.4 | NP_058033.2 | 3 - 4 | 564 | 74 | |
GenBank mRNA | AK032170.1 | - | 3 - 4 | 565 | 74 | |
AK149981.1 | - | 3 - 4 | 524 | 74 | ||
BC115521.1 | - | 3 - 4 | 560 | 40101205 | 74 | |
BC115522.1 | - | 3 - 4 | 562 | 40101207 | 74 | |
D88769.1 | - | 3 - 4 | 470 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |