Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271584.1 | NP_001258513.1 | 3 - 4 | 381 | 75 | |
NM_001271585.1 | NP_001258514.1 | 3 - 4 | 408 | 75 | ||
NM_016775.3 | NP_058055.1 | 2 - 3 | 328 | 75 | ||
NR_073369.1 | 2 - 3 | 328 | 75 | |||
GenBank mRNA | AF032115.1 | - | 1 - 2 | 104 | 75 | |
AK007879.1 | - | 2 - 3 | 271 | 75 | ||
AK029006.1 | - | 2 - 3 | 266 | 75 | ||
AK032373.1 | - | 2 - 3 | 260 | 75 | ||
AK083584.1 | - | 2 - 3 | 289 | 75 | ||
BC012268.1 | - | 3 - 4 | 3500124 | 75 | ||
BC018231.1 | - | 2 - 3 | 255 | 75 | ||
BY101584.1 | - | 3 - 4 | 316 | 75 | ||
CB249513.1 | - | 3 - 4 | 295 | 75 | ||
CN459638.1 | - | 2 - 3 | 144 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |