Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016963.2 | NP_058659.1 | 5 - 6 | 694 | 79 | |
GenBank mRNA | AF177172.1 | - | 5 - 6 | 682 | 79 | |
AF237630.1 | - | 5 - 6 | 608 | 79 | ||
AK050372.1 | - | 5 - 6 | 696 | 79 | ||
AK084463.1 | - | 2 - 3 | 216 | 79 | ||
AK167723.1 | - | 5 - 6 | 692 | 79 | ||
BC082595.1 | - | 5 - 6 | 686 | 30645500 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |