Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289514.1 | NP_001276443.1 | 9 - 10 | 1372 | 96 | |
NM_019636.3 | NP_062610.2 | 10 - 11 | 1953 | 96 | ||
GenBank mRNA | AK122445.1 | - | 10 - 11 | 2762 | 96 | |
AK135665.1 | - | 9 - 10 | 1544 | 96 | ||
AK135892.1 | - | 9 - 10 | 1637 | 96 | ||
AK148356.1 | - | 9 - 10 | 1329 | 96 | ||
AK154591.1 | - | 11 - 12 | 1989 | 96 | ||
AK159256.1 | - | 10 - 11 | 1949 | 96 | ||
AK164923.1 | - | 6 - 7 | 657 | 96 | ||
BC059824.1 | - | 11 - 12 | 2318 | 96 | ||
BC117931.1 | - | 9 - 10 | 1621 | 40097712 | 96 | |
U33005.1 | - | 10 - 11 | 1909 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |