Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pias1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019663.3 | NP_062637.2 | 5 - 6 | 697 | 91 | |
GenBank mRNA | AF077950.1 | - | 5 - 6 | 689 | 91 | |
AK075708.1 | - | 5 - 6 | 698 | 91 | ||
AK153871.1 | - | 5 - 6 | 702 | 91 | ||
BC051417.1 | - | 5 - 6 | 694 | 6515027 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |