Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pias1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019663.3 | NP_062637.2 | 10 - 11 | 1310 | 69 | |
GenBank mRNA | AF077950.1 | - | 10 - 11 | 1302 | 69 | |
AK013753.1 | - | 6 - 7 | 644 | 69 | ||
AK075708.1 | - | 10 - 11 | 1311 | 69 | ||
AK153871.1 | - | 10 - 11 | 1315 | 69 | ||
BC051417.1 | - | 10 - 11 | 1307 | 6515027 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |