Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019670.1 | NP_062644.1 | 4 - 5 | 430 | 107 | |
GenBank mRNA | AF094519.1 | - | 4 - 5 | 430 | 107 | |
AK137073.1 | - | 3 - 4 | 519 | 107 | ||
AK138694.1 | - | 4 - 5 | 460 | 107 | ||
AK161737.1 | - | 3 - 4 | 448 | 107 | ||
AK166882.1 | - | 4 - 5 | 486 | 107 | ||
AK220493.1 | - | 4 - 5 | 494 | 107 | ||
BC003250.1 | - | 3 - 4 | 464 | 107 | ||
BC085191.1 | - | 4 - 5 | 540 | 30693873 | 107 | |
BC086779.1 | - | 4 - 5 | 505 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |