Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29050153 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29050150 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29050149 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29050152 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P121198 |
AF205936.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P121198 |
NM_019968.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P121198 |
AF205936.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P121198 |
NM_019968.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
719496 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
790377 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
529787 |
AK085572.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
529787 |
BC025462.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
529787 |
BC027836.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
529787 |
NM_145946.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
601114 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
7122 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
145539 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
67538 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P109441 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P121198 |
AF205936.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P121198 |
NM_019968.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P121198 |
AF205936.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P121198 |
NM_019968.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P271591 |
AK090305.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P121198 |
AF205936.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P121198 |
NM_019968.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P121198 |
AF205936.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P121198 |
NM_019968.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29050155 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29050158 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29050153 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29050166 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29050160 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29050150 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29050165 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29050168 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29050152 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29050151 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29050163 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P109441 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07880530 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P121198 |
AF205936.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P121198 |
NM_019968.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P121198 |
AF205936.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P121198 |
NM_019968.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P271591 |
AK090305.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P109400 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P137111 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P109441 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10966824 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29050152 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29050153 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29050149 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29050150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050260 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050198 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050173 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050103 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050245 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050164 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050190 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050230 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050186 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050219 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050189 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050166 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050188 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050251 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050242 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050258 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050137 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050208 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050120 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050232 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050192 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050179 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050187 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050153 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050211 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050158 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050214 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050238 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050250 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050222 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050202 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050169 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050177 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050261 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050143 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050170 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050163 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050200 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050172 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050174 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050147 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050255 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050160 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050247 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050221 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050203 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050180 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050194 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050224 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050168 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050213 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050234 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050215 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050155 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050252 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29050217 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10966825 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050195 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050170 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050181 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050168 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10609191 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10966828 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050139 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050143 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10846152 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10798271 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050130 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050202 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10798270 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050188 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10609192 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050183 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050190 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050179 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050129 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050137 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050166 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050146 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050174 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10995551 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10716185 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050131 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10955889 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11113305 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050198 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050165 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050150 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050173 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050128 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10995546 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050163 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050136 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050192 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10484233 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11113300 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050164 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050186 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11027880 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050172 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050187 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050153 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10737998 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050158 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050194 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10484232 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11002579 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050155 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29050177 |
- |
- |