Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199060.1 | NP_001185989.1 | 9 - 10 | 1284 | 106 | |
NM_001199061.1 | NP_001185990.1 | 9 - 10 | 1016 | 106 | ||
NM_021312.5 | NP_067287.1 | 8 - 9 | 1106 | 106 | ||
GenBank mRNA | AB041608.1 | - | 9 - 10 | 1285 | 106 | |
AK007500.1 | - | 8 - 9 | 883 | 106 | ||
AK016611.1 | - | 9 - 10 | 960 | 106 | ||
AK031436.1 | - | 8 - 9 | 869 | 106 | ||
AK149085.1 | - | 8 - 9 | 788 | 106 | ||
AK161299.1 | - | 9 - 10 | 1029 | 106 | ||
AY059431.1 | - | 8 - 9 | 797 | 106 | ||
AY059432.1 | - | 1 - 2 | 102 | 106 | ||
BC004748.1 | - | 8 - 9 | 940 | 3583955 | 106 | |
BC052386.1 | - | 8 - 9 | 1077 | 6418498 | 106 | |
BC096651.1 | - | 8 - 9 | 851 | 1514703 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |